Anti-BAX Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48015-100UL A48015-50UL
77713-923EA 529.22 CAD
77713-923 77713-924
Anti-BAX Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: BAX
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR