Anti-ART5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47994-100UL A47994-50UL
77713-893EA 529.22 CAD
77713-893 77713-894
Anti-ART5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: ART5
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR