Anti-CNP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47980-50UG A47980-100UG
77713-871EA 360.84 CAD
77713-871 77713-870
Anti-CNP Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: CNP
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR