Anti-ICAM3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47925-50UL A47925-100UL
77713-777EA 360.84 CAD
77713-777 77713-776
Anti-ICAM3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: ICAM3
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR