Anti-CALCA Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47916-100UG A47916-50UG
77705-357EA 400.93 CAD
77705-357 77713-763
Anti-CALCA Rabbit Polyclonal Antibody
Antibodies
WB:1:500-2000, ELISA:1:10000-20000

Type:
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR