Anti-ERVW-1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47886-50UL A47886-100UL
77713-713EA 360.84 CAD
77713-713 77713-712
Anti-ERVW-1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: ERVW-1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol ERVW-1
Environmentally Preferable
Antigen synonyms Enverin|gp50)|Syncytin) [Cleaved into: Surface protein|gp24)]|Transmembrane protein|ERVW-1|HERV-W_7q21.2 provirus ancestral Env polyprotein|ERVWE1|HERV-W envelope protein|Syncytin-1|TM|Env-W|Envelope polyprotein gPr73|HERV-7q Envelope protein|Endogenous retrovirus group W member 1|SU
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID Q9UQF0
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide of Human ERVW-1
Form Liquid
Purification Antigen affinity purified
Storage buffer PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR