Anti-SPR Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47870-50UL A47870-100UL
77713-686EA 360.84 CAD
77713-686 77713-685
Anti-SPR Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SPR
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR