Anti-EPAS1 (acetyl-K385) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47853-100UG A47853-50UG
77705-353EA 400.93 CAD
77705-353 77713-657
Anti-EPAS1 (acetyl-K385) Rabbit Polyclonal Antibody
Antibodies
WB:1:500-2000, ELISA:1:10000-20000

Type:
Antigen: EPAS1 (acetyl-K385)
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR