Anti-PPP2R3A Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47791-50UG A47791-100UG
77713-560EA 360.84 CAD
77713-560 77713-559
Anti-PPP2R3A Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PPP2R3A
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR