Anti-ROR2 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47786-50UG A47786-100UG
77713-550EA 360.84 CAD
77713-550 77713-549
Anti-ROR2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ROR2
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR