Anti-VWCE Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47760-50UL A47760-100UL
77713-502EA 360.84 CAD
77713-502 77713-501
Anti-VWCE Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: VWCE
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR