Anti-ZBTB16 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47746-100UG A47746-50UG
77713-475EA 529.22 CAD
77713-475 77713-476
Anti-ZBTB16 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ZBTB16
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR