Anti-ASNS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47741-50UL A47741-100UL
77713-467EA 360.84 CAD
77713-467 77713-466
Anti-ASNS Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: ASNS
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR