Anti-tbx16 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47731-100UG A47731-50UG
77713-447EA 529.22 CAD
77713-447 77713-448
Anti-tbx16 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: tbx16
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR