Anti-dharma Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47718-50UG A47718-100UG
77713-423EA 360.84 CAD
77713-423 77713-422
Anti-dharma Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: dharma
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR