Anti-egfra Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47696-100UG A47696-50UG
77713-382EA 529.22 CAD
77713-382 77713-383
Anti-egfra Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: egfra
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR