Anti-nkx2.5 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47687-50UG A47687-100UG
77713-367EA 360.84 CAD
77713-367 77713-366
Anti-nkx2.5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: nkx2.5
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR