Anti-RAB41 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47676-50UL A47676-100UL
77713-347EA 360.84 CAD
77713-347 77713-346
Anti-RAB41 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RAB41
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR