Anti-PAX5 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47625-100UG A47625-50UG
77713-264EA 529.22 CAD
77713-264 77713-265
Anti-PAX5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PAX5
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR