Anti-SRGAP2C Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47571-100UG A47571-50UG
77713-175EA 529.22 CAD
77713-175 77713-176
Anti-SRGAP2C Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SRGAP2C
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR