Anti-atpB Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47554-50UG A47554-100UG
77713-146EA 360.84 CAD
77713-146 77713-145
Anti-atpB Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: atpB
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR