Anti-AKR1B1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47540-50UL A47540-100UL
77713-124EA 360.84 CAD
77713-124 77713-123
Anti-AKR1B1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200

Type:
Antigen: AKR1B1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR