Anti-TERT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47465-50UL A47465-100UL
77712-999EA 360.84 CAD
77712-999 77712-998
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR