Anti-KLHL1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47463-50UL A47463-100UL
77712-996EA 360.84 CAD
77712-996 77712-995
Anti-KLHL1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: KLHL1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR