Anti-OLE9 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47460-50UG A47460-100UG
77712-991EA 360.84 CAD
77712-991 77712-990
Anti-OLE9 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: OLE9
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Olea europaea
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR