Anti-OLE9 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A47458-100UG A47458-50UG
77712-986EA 529.22 CAD
77712-986 77712-987
Anti-OLE9 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: OLE9
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Olea europaea
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR