Anti-BETV4 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47445-50UG A47445-100UG
77712-966EA 360.84 CAD
77712-966 77712-965
Anti-BETV4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: BETV4
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Betula pendula
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR