Anti-Major pollen allergen Aln g 1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A47436-100UG A47436-50UG
77712-948EA 529.22 CAD
77712-948 77712-949
Anti-Major pollen allergen Aln g 1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Major pollen allergen Aln g 1
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Alnus glutinosa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR