Anti-Allergen Bla g 4 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47424-50UG A47424-100UG
77712-932EA 360.84 CAD
77712-932 77712-931
Anti-Allergen Bla g 4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Allergen Bla g 4
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Blattella germanica
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR