Anti-AGXT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47413-100UL A47413-50UL
77712-910EA 529.22 CAD
77712-910 77712-911
Anti-AGXT Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: AGXT
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR