Anti-Allergen Fel d 4 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47412-100UG A47412-50UG
77712-908EA 529.22 CAD
77712-908 77712-909
Anti-Allergen Fel d 4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Allergen Fel d 4
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR