Anti-NLRP10 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47392-50UL A47392-100UL
77712-872EA 360.84 CAD
77712-872 77712-871
Anti-NLRP10 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: NLRP10
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR