Anti-MFSD2A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47375-50UL A47375-100UL
77712-843EA 360.84 CAD
77712-843 77712-842
Anti-MFSD2A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: MFSD2A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR