Anti-BCR Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47372-100UL A47372-50UL
77712-836EA 529.22 CAD
77712-836 77712-837
Anti-BCR Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: BCR
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR