Anti-WNT2B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47371-50UL A47371-100UL
77712-835EA 360.84 CAD
77712-835 77712-834
Anti-WNT2B Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:10-1:50

Type:
Antigen: WNT2B
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR