Anti-TH Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47349-100UL A47349-50UL
77712-795EA 529.22 CAD
77712-795 77712-796
Anti-TH Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TH
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR