Anti-BAAT Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47339-100UL A47339-50UL
77712-778EA 529.22 CAD
77712-778 77712-779
Anti-BAAT Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: BAAT
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR