Anti-WNT10B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47256-100UL A47256-50UL
77712-639EA 529.22 CAD
77712-639 77712-640
Anti-WNT10B Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:10-1:50

Type:
Antigen: WNT10B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR