Anti-Tubulin beta-1 chain Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A47057-50UG A47057-100UG
77712-300EA 360.84 CAD
77712-300 77712-299
Anti-Tubulin beta-1 chain Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Tubulin beta-1 chain
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gallus gallus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR