Anti-Tubulin beta-1 chain Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47058-100UG A47058-50UG
77712-301EA 529.22 CAD
77712-301 77712-302
Anti-Tubulin beta-1 chain Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Tubulin beta-1 chain
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gallus gallus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR