Anti-Streptavidin Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A47025-50UG A47025-100UG
77712-247EA 360.84 CAD
77712-247 77712-246
Anti-Streptavidin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. avidinii
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR