Anti-SFTPB Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46964-50UG A46964-100UG
77712-134EA 360.84 CAD
77712-134 77712-133
Anti-SFTPB Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SFTPB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR