Anti-SERPINB6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46741-50UL A46741-100UL
77711-744EA 360.84 CAD
77711-744 77711-743
Anti-SERPINB6 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: SERPINB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR