Anti-RPA3 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46322-50UG A46322-100UG
77710-990EA 360.84 CAD
77710-990 77710-989
Anti-RPA3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: RPA3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR