Anti-rho Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A46447-100UG A46447-50UG
77711-213EA 529.22 CAD
77711-213 77711-214
Anti-rho Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: rho
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR