Anti-rho Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A46453-50UG A46453-100UG
77711-226EA 360.84 CAD
77711-226 77711-225
Anti-rho Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: rho
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR