Anti-rho Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A46448-100UG A46448-50UG
77711-215EA 529.22 CAD
77711-215 77711-216
Anti-rho Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: rho
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR