Anti-rho Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46449-100UG A46449-50UG
77711-217EA 529.22 CAD
77711-217 77711-218
Anti-rho Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: rho
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR