Anti-rho Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46446-50UG A46446-100UG
77711-212EA 360.84 CAD
77711-212 77711-211
Anti-rho Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: rho
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR