Anti-recQ Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A46934-50UG A46934-100UG
77712-083EA 360.84 CAD
77712-083 77712-082
Anti-recQ Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: recQ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR